1,1,1,2,2-Pentafluoropentan-3-ol
Supplier: Apollo Scientific
Warning
PC1792-1G PC1792-5G
APOSPC1792-1GEA 236 EUR
APOSPC1792-1G APOSPC1792-5G
1,1,1,2,2-Pentafluoropentan-3-ol
1,1,1,2,2-Pentafluoropentan-3-ol
Formula: C₅H₇F₅O
MW: 178.1 g/mol
Boiling Pt: 97…98 °C (760 mm Hg)
Density: 1.329 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784220
CAS Number: 378-71-2
UN: 1993
ADR: 3,III