2-Fluoro-[1,1-biphenyl]-4-magnesiumbromide 0.5M in THF
Supplier: Apollo Scientific
PC1844-100ML PC1844-50ML
APOSPC1844-100MLEA 332 EUR
APOSPC1844-100ML APOSPC1844-50ML
2-Fluoro-[1,1-biphenyl]-4-magnesiumbromide 0.5M in THF
2-Fluoro-[1,1-biphenyl]-4-magnesiumbromide
Formula: C₁₂H₈BrFMg
MW: 275.399 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07698745
CAS Number: 76699-46-2