Zinc(II) perfluorobutane-1-sulphonate
Supplier: Apollo Scientific
Warning
Synonyms: Zinc(II) 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulphonate, Zinc(II) nonaflate
PC1941-5G PC1941-25G
APOSPC1941-5GEA 113 EUR
APOSPC1941-5G APOSPC1941-25G
Zinc(II) perfluorobutane-1-sulphonate
Zinc(II) perfluorobutane-1-sulphonate
Formula: Zn(C₄F₉O₃S)₂
MW: 663.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094139
CAS Number: 502457-69-4