4-Acetyl-2,2-difluoro-1,3-benzodioxole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,2-Difluoro-1,3-benzodioxol-4-yl)ethan-1-one, 1-(2,2-Difluoro-1,3-benzodioxol-4-yl)ethanone, 2',3'-(Difluoromethylenedioxy)acetophenone
PC1954-1G PC1954-250MG
APOSPC1954-1GEA 261 EUR
APOSPC1954-1G APOSPC1954-250MG
4-Acetyl-2,2-difluoro-1,3-benzodioxole 98%
4-Acetyl-2,2-difluoro-1,3-benzodioxole
Formula: C₉H₆F₂O₃
MW: 200.139 g/mol
Boiling Pt: 87…89 °C (12 mm Hg)
Melting Pt: 32…33 °C
Storage Temperature: Ambient
MDL Number: MFCD08458053
CAS Number: 126120-83-0