5-Acetyl-2,2-difluoro-1,3-benzodioxole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,2-Difluoro-1,3-benzodioxol-5-yl)ethan-1-one, 3',4'-(Difluoromethylenedioxy)acetophenone
PC1955-1G PC1955-250MG
APOSPC1955-1GEA 493 EUR
APOSPC1955-1G APOSPC1955-250MG
5-Acetyl-2,2-difluoro-1,3-benzodioxole 98%
5-Acetyl-2,2-difluoro-1,3-benzodioxole
Formula: C₉H₆F₂O₃
MW: 200.139 g/mol
Boiling Pt: 97…99 °C (7.5 mm Hg)
Melting Pt: 31…32 °C
Storage Temperature: Ambient
MDL Number: MFCD07777109
CAS Number: 136593-45-8