5-Bromo-3-iodo-6-(trifluoromethyl)-1H-indole
Supplier: Apollo Scientific
PC200036-1G
PC200036-500MG
APOSPC200036-1GEA
527
EUR
APOSPC200036-1G
APOSPC200036-500MG
5-Bromo-3-iodo-6-(trifluoromethyl)-1H-indole
5-Bromo-3-iodo-6-(trifluoromethyl)-1H-indole
Formula:
C₉H₄BrF₃IN MW: 389.938 g/mol Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD22689162 CAS Number: 1420537-61-6 |