Methyl 3-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-6-carboxylate
Supplier: Apollo Scientific
PC200075-1G PC200075-500MG
APOSPC200075-1GEA 326 EUR
APOSPC200075-1G APOSPC200075-500MG
Methyl 3-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-6-carboxylate
Methyl 3-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-6-carboxylate
Formula: C₁₀H₆F₃IN₂O₂
Melting Pt: 158…161 °C
Storage Temperature: Ambient
MDL Number: MFCD24447583
CAS Number: 1432895-08-3