1-(1-{[3-(Trifluoromethyl)phenyl]methyl}-1H-1,2,3-triazol-4-yl)ethan-1-ol
Supplier: Apollo Scientific
PC200080-1G PC200080-500MG
APOSPC200080-1GEA 187 EUR
APOSPC200080-1G APOSPC200080-500MG
1-(1-{[3-(Trifluoromethyl)phenyl]methyl}-1H-1,2,3-triazol-4-yl)ethan-1-ol
1-(1-{[3-(Trifluoromethyl)phenyl]methyl}-1H-1,2,3-triazol-4-yl)ethan-1-ol
Formula: C₁₂H₁₂F₃N₃O
MW: 271.238 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD25121137
CAS Number: 1443291-26-6