Potassium trifluoro(phenylethynyl)borate 95%
Supplier: Apollo Scientific
Warning
PC200094-1G PC200094-500MG PC200094-5G
APOSPC200094-1GEA 106 EUR
APOSPC200094-1G APOSPC200094-500MG APOSPC200094-5G
Potassium trifluoro(phenylethynyl)borate 95%
Potassium trifluoro(phenylethynyl)borate
Formula: C₈H₅BF₃·K
Boiling Pt: >125 °C
Melting Pt: 250…255 °C
Density: >1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD11052662
CAS Number: 485338-93-0