(1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl)methanol 95%
Supplier: Apollo Scientific
PC200099-500MG PC200099-1G
APOSPC200099-500MGEA 236 EUR
APOSPC200099-500MG APOSPC200099-1G
(1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl)methanol 95%
(1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl)methanol
Formula: C₆H₇F₃N₂O
MW: 180.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18262424
CAS Number: 1260659-13-9