(3'-(Trifluoromethyl)-[1,1'-biphenyl]-3-yl)methanol
Supplier: Apollo Scientific
PC200129-1G PC200129-500MG PC200129-5G
APOSPC200129-1GEA 180 EUR
APOSPC200129-1G APOSPC200129-500MG APOSPC200129-5G
(3'-(Trifluoromethyl)-[1,1'-biphenyl]-3-yl)methanol
(3'-(Trifluoromethyl)-[1,1'-biphenyl]-3-yl)methanol
Formula: C₁₄H₁₁F₃O
MW: 252.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05980218
CAS Number: 773872-61-0