2-Bromo-1-(5-fluoro-2-methoxyphenyl)ethan-1-one
Supplier: Apollo Scientific
Synonyms: 2-Bromo-1-(5-fluoro-2-methoxyphenyl)ethanone
PC200563-1G PC200563-25G PC200563-5G
APOSPC200563-1GEA 43.7 EUR
APOSPC200563-1G APOSPC200563-25G APOSPC200563-5G
2-Bromo-1-(5-fluoro-2-methoxyphenyl)ethan-1-one
2-Bromo-1-(5-fluoro-2-methoxyphenyl)ethan-1-one
Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD03425169
CAS Number: 343-04-4