Ethyl 2-amino-4,6-difluorobenzoate 95+%
Supplier: Apollo Scientific
PC201028-100MG PC201028-1G PC201028-250MG
APOSPC201028-100MGEA 79.9 EUR
APOSPC201028-100MG APOSPC201028-1G APOSPC201028-250MG
Ethyl 2-amino-4,6-difluorobenzoate 95+%
Ethyl 2-amino-4,6-difluorobenzoate
Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12159820
CAS Number: 1147107-15-0