6-Chloro-4-(trifluoromethyl)picolinonitrile 97%
Supplier: Apollo Scientific
PC201034-1G PC201034-250MG
APOSPC201034-1GEA 763 EUR
APOSPC201034-1G APOSPC201034-250MG
6-Chloro-4-(trifluoromethyl)picolinonitrile 97%
6-Chloro-4-(trifluoromethyl)picolinonitrile
Formula: C₇H₂ClF₃N₂
MW: 206.552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00106773
CAS Number: 1156542-25-4