5-Chloro-3-(trifluoromethyl)pyridin-2-ol 97%
Supplier: Apollo Scientific
PC201071-1G
APOSPC201071-1GEA 95.7 EUR
APOSPC201071-1G
5-Chloro-3-(trifluoromethyl)pyridin-2-ol 97%
5-Chloro-3-(trifluoromethyl)pyridin-2-ol
Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20275098
CAS Number: 1214342-70-7