(4-(Trifluoromethyl)pyrimidin-2-yl)methanol 97%
Supplier: Apollo Scientific
PC201086-100MG PC201086-1G PC201086-250MG
APOSPC201086-100MGEA 122 EUR
APOSPC201086-100MG APOSPC201086-1G APOSPC201086-250MG
(4-(Trifluoromethyl)pyrimidin-2-yl)methanol 97%
(4-(Trifluoromethyl)pyrimidin-2-yl)methanol
Formula: C₆H₅F₃N₂O
MW: 178.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16987898
CAS Number: 1240594-67-5