6-Chloro-2-(trifluoromethyl)nicotinonitrile 98%
Supplier: Apollo Scientific
Synonyms: 6-Chloro-2-(trifluoromethyl)-3-pyridinecarbonitrile
PC201091-100MG PC201091-1G PC201091-250MG
APOSPC201091-100MGEA 43.5 EUR
APOSPC201091-100MG APOSPC201091-1G APOSPC201091-250MG
6-Chloro-2-(trifluoromethyl)nicotinonitrile 98%
6-Chloro-2-(trifluoromethyl)nicotinonitrile
Formula: C₇H₂ClF₃N₂
MW: 206.552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848371
CAS Number: 1245913-20-5