7-Fluoroimidazo[1,2-a]pyridine 95%
Supplier: Apollo Scientific
Warning
PC201102-1G PC201102-250MG PC201102-5G
APOSPC201102-1GEA 62.5 EUR
APOSPC201102-1G APOSPC201102-250MG APOSPC201102-5G
7-Fluoroimidazo[1,2-a]pyridine 95%
7-Fluoroimidazo[1,2-a]pyridine
Formula: C₇H₅FN₂
MW: 136.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11976201
CAS Number: 1260903-17-0