1,5-Difluoro-3-methyl-2-nitrobenzene 98%
Supplier: Apollo Scientific
PC201159-1G PC201159-250MG PC201159-5G
APOSPC201159-1GEA 88.6 EUR
APOSPC201159-1G APOSPC201159-250MG APOSPC201159-5G
1,5-Difluoro-3-methyl-2-nitrobenzene 98%
1,5-Difluoro-3-methyl-2-nitrobenzene
Formula: C₇H₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD21603715
CAS Number: 1616526-80-7