3-(2,2,2-Trifluoroacetyl)benzoic acid 95+%
Supplier: Apollo Scientific
Warning
PC201194-100MG PC201194-250MG
APOSPC201194-100MGEA 470 EUR
APOSPC201194-100MG APOSPC201194-250MG
3-(2,2,2-Trifluoroacetyl)benzoic acid 95+%
3-(2,2,2-Trifluoroacetyl)benzoic acid
Formula: C₉H₅F₃O₃
Melting Pt: 94…95 °C
Storage Temperature: Ambient
MDL Number: MFCD13173376
CAS Number: 213598-05-1