5-Bromo-3-fluoro-2-methoxyaniline 95+%
Supplier: Apollo Scientific
PC201209-1G PC201209-250MG
APOSPC201209-1GEA 253 EUR
APOSPC201209-1G APOSPC201209-250MG
5-Bromo-3-fluoro-2-methoxyaniline 95+%
5-Bromo-3-fluoro-2-methoxyaniline
Formula: C₇H₇BrFNO
MW: 220.039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847600
CAS Number: 239122-51-1