3-(Difluoromethoxy)-4-methylaniline 98%
Supplier: Apollo Scientific
PC201218-1G PC201218-250MG
APOSPC201218-1GEA 207 EUR
APOSPC201218-1G APOSPC201218-250MG
3-(Difluoromethoxy)-4-methylaniline 98%
3-(Difluoromethoxy)-4-methylaniline
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025429
CAS Number: 264194-31-2