(R)-1-(3,4-Difluorophenyl)ethanamine 95+%
Supplier: Apollo Scientific
PC201230-100MG PC201230-1G PC201230-250MG
APOSPC201230-100MGEA 104 EUR
APOSPC201230-100MG APOSPC201230-1G APOSPC201230-250MG
(R)-1-(3,4-Difluorophenyl)ethanamine 95+%
(R)-1-(3,4-Difluorophenyl)ethanamine
Formula: C₈H₉F₂N·ClH
MW: 193.622 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06761760
CAS Number: 321318-15-4