2-(4,4-Difluoropiperidin-1-yl)ethanamine 95+%
Supplier: Apollo Scientific
PC201267-100MG PC201267-250MG PC201267-25MG
APOSPC201267-100MGEA 303 EUR
APOSPC201267-100MG APOSPC201267-250MG APOSPC201267-25MG
2-(4,4-Difluoropiperidin-1-yl)ethanamine 95+%
2-(4,4-Difluoropiperidin-1-yl)ethanamine
Formula: C₇H₁₄F₂N₂
MW: 164.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461787
CAS Number: 605659-03-8