(5-Fluoro-2-methoxypyridin-3-yl)methanol 98%
Supplier: Apollo Scientific
PC201339-100MG PC201339-250MG PC201339-1G
APOSPC201339-100MGEA 43.9 EUR
APOSPC201339-100MG APOSPC201339-250MG APOSPC201339-1G
(5-Fluoro-2-methoxypyridin-3-yl)methanol 98%
(5-Fluoro-2-methoxypyridin-3-yl)methanol
Formula: C₇H₈FNO₂
MW: 157.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972415
CAS Number: 874822-98-7