2-Chloro-6-(trifluoromethyl)thiazolo[4,5-b]pyridine 98%
Supplier: Apollo Scientific
PC201354-100MG PC201354-250MG PC201354-1G
APOSPC201354-100MGEA 104 EUR
APOSPC201354-100MG APOSPC201354-250MG APOSPC201354-1G
2-Chloro-6-(trifluoromethyl)thiazolo[4,5-b]pyridine 98%
2-Chloro-6-(trifluoromethyl)thiazolo[4,5-b]pyridine
Formula: C₇H₂ClF₃N₂S
MW: 238.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193434
CAS Number: 884860-62-2