5-Fluoro-1H-pyrrolo[3,2-b]pyridine 98%
Supplier: Apollo Scientific
PC201361-100MG PC201361-250MG PC201361-1G
APOSPC201361-100MGEA 253 EUR
APOSPC201361-100MG APOSPC201361-250MG APOSPC201361-1G
5-Fluoro-1H-pyrrolo[3,2-b]pyridine 98%
5-Fluoro-1H-pyrrolo[3,2-b]pyridine
Formula: C₇H₅FN₂
MW: 136.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062552
CAS Number: 887570-96-9