3,4-Difluoro-5-hydroxybenzaldehyde 97%
Supplier: Apollo Scientific
PC201362-1G PC201362-5G
APOSPC201362-1GEA 69.6 EUR
APOSPC201362-1G APOSPC201362-5G
3,4-Difluoro-5-hydroxybenzaldehyde 97%
3,4-Difluoro-5-hydroxybenzaldehyde
Formula: C₇H₄F₂O₂
MW: 158.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972451
CAS Number: 887584-91-0