6-Bromo-5-fluoroindolin-2-one 96%
Supplier: Apollo Scientific
PC201365-100MG PC201365-250MG PC201365-1G
APOSPC201365-100MGEA 111 EUR
APOSPC201365-100MG APOSPC201365-250MG APOSPC201365-1G
6-Bromo-5-fluoroindolin-2-one 96%
6-Bromo-5-fluoroindolin-2-one
Formula: C₈H₅BrFNO
MW: 230.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16250437
CAS Number: 893620-44-5