6-(Difluoromethyl)pyridin-3-amine 97%
Supplier: Apollo Scientific
PC201370-250MG PC201370-1G
APOSPC201370-250MGEA 683 EUR
APOSPC201370-250MG APOSPC201370-1G
6-(Difluoromethyl)pyridin-3-amine 97%
6-(Difluoromethyl)pyridin-3-amine
Formula: C₆H₆F₂N₂
MW: 144.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384489
CAS Number: 913090-41-2