3-Chloro-4-fluoro-5-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
PC201372-250MG PC201372-1G PC201372-5G
APOSPC201372-250MGEA 63.8 EUR
APOSPC201372-250MG APOSPC201372-1G APOSPC201372-5G
3-Chloro-4-fluoro-5-(trifluoromethyl)aniline 98%
3-Chloro-4-fluoro-5-(trifluoromethyl)aniline
Formula: C₇H₄ClF₄N
MW: 213.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033155
CAS Number: 914225-61-9