5-Methoxy-4'-(trifluoromethyl)valerophenone
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methoxy-1-(4-(trifluoromethyl)phenyl)pentan-1-one, 4-(5-Methoxypentanoyl)benzotrifluoride, Fluvoxamine Maleate EP Impurity D, 5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one
PC2055-1G PC2055-25G PC2055-5G
APOSPC2055-1GEA 34.8 EUR
APOSPC2055-1G APOSPC2055-25G APOSPC2055-5G
5-Methoxy-4'-(trifluoromethyl)valerophenone
5-Methoxy-4'-(trifluoromethyl)valerophenone
Formula: C₁₃H₁₅F₃O₂
MW: 260.252 g/mol
Melting Pt: 42…43 °C
Storage Temperature: Ambient
MDL Number: MFCD06658173
CAS Number: 61718-80-7