N-(4-Fluorophenyl)-3-methyl-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
Supplier: Apollo Scientific
PC210050-1G
APOSPC210050-1GEA 780 EUR
APOSPC210050-1G
N-(4-Fluorophenyl)-3-methyl-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
N-(4-Fluorophenyl)-3-methyl-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
Formula: C₁₃H₁₁FN₄O₂S
MW: 306.315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18804108
CAS Number: 1291487-08-5