N-(2H-1,3-Benzodioxol-5-yl)-2,2,2-trifluoroacetamide
Supplier: Apollo Scientific
Warning
PC210075-1G PC210075-500MG
APOSPC210075-1GEA 297 EUR
APOSPC210075-1G APOSPC210075-500MG
N-(2H-1,3-Benzodioxol-5-yl)-2,2,2-trifluoroacetamide
N-(2H-1,3-Benzodioxol-5-yl)-2,2,2-trifluoroacetamide
Formula: C₉H₆F₃NO₃
MW: 233.144 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00455931
CAS Number: 85575-56-0