1,5-Dichloro-2-[(3-fluorophenyl)methoxy]-4-nitrobenzene
Supplier: Apollo Scientific
PC210082-1G PC210082-500MG
APOSPC210082-1GEA 297 EUR
APOSPC210082-1G APOSPC210082-500MG
1,5-Dichloro-2-[(3-fluorophenyl)methoxy]-4-nitrobenzene
1,5-Dichloro-2-[(3-fluorophenyl)methoxy]-4-nitrobenzene
Formula: C₁₃H₈Cl₂FNO₃
MW: 316.112 g/mol
Melting Pt: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD28506331
CAS Number: 1708126-05-9