2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]sulphanyl}ethan-1-amine hydrochloride
Supplier: Apollo Scientific
PC210084-1G PC210084-500MG
APOSPC210084-1GEA 223 EUR
APOSPC210084-1G APOSPC210084-500MG
2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]sulphanyl}ethan-1-amine hydrochloride
2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]sulphanyl}ethan-1-amine hydrochloride
Formula: C₈H₈ClF₃N₂S·ClH
MW: 293.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08447118
CAS Number: 1177351-21-1