3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine ≥98%
Supplier: Apollo Scientific
Warning
PC210142-100MG PC210142-1G PC210142-250MG
APOSPC210142-100MGEA 0 EUR
APOSPC210142-100MG APOSPC210142-1G APOSPC210142-250MG
3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine ≥98%
3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Formula: C₈H₇F₃N₂
MW: 188.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024501
CAS Number: 1150618-34-0