5,6-Difluoro-2,3-dihydro-1H-isoindol-1-one ≥98%
Supplier: Apollo Scientific
Synonyms: 5,6-Difluoro-2,3-dihydro-1h-isoindol-1-one
PC210145-100MG PC210145-1G PC210145-250MG
APOSPC210145-100MGEA 190 EUR
APOSPC210145-100MG APOSPC210145-1G APOSPC210145-250MG
5,6-Difluoro-2,3-dihydro-1H-isoindol-1-one ≥98%
5,6-Difluoro-2,3-dihydro-1H-isoindol-1-one
Formula: C₈H₅F₂NO
MW: 169.128 g/mol
MDL Number: MFCD24617103
CAS Number: 1192040-50-8