2,3-Bis(trifluoromethyl)nitrobenzene 98%
Supplier: Apollo Scientific
Warning
PC2287-1G PC2287-250MG
APOSPC2287-1GEA 636 EUR
APOSPC2287-1G APOSPC2287-250MG
2,3-Bis(trifluoromethyl)nitrobenzene 98%
2,3-Bis(trifluoromethyl)nitrobenzene
Formula: C₈H₃F₆NO₂
MW: 259.105 g/mol
Boiling Pt: 223…225 °C (760 mm Hg)
Melting Pt: 26…29 °C
Storage Temperature: Ambient
MDL Number: MFCD04972858
CAS Number: 1978-06-9