1-(4-Chlorophenyl)-3-(2,6-difluorobenzoyl)urea 95%
Supplier: Apollo Scientific
Synonyms: Diflubenzuron
PC2299-100G PC2299-25G PC2299-5G
APOSPC2299-100GEA 447 EUR
APOSPC2299-100G APOSPC2299-25G APOSPC2299-5G
1-(4-Chlorophenyl)-3-(2,6-difluorobenzoyl)urea 95%
1-(4-Chlorophenyl)-3-(2,6-difluorobenzoyl)urea
Formula: C₁₄H₉ClF₂N₂O₂
MW: 310.683 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055343
CAS Number: 35367-38-5