3-(Difluoromethoxy)benzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
PC2382-5G PC2382-25G
APOSPC2382-5GEA 244 EUR
APOSPC2382-5G APOSPC2382-25G
3-(Difluoromethoxy)benzenesulphonyl chloride
3-(Difluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₅ClF₂O₃S
MW: 242.628 g/mol
Boiling Pt: 233…234 °C
Density: 1.509 g/cm³ (25 °C)
Flash Pt: 97.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03094157
CAS Number: 351003-38-8
UN: 3265
ADR: 8 II