4-Fluoro-2-(methoxymethyl)bromobenzene 98%
Supplier: Apollo Scientific
PC2421-1G PC2421-250MG
APOSPC2421-1GEA 667 EUR
APOSPC2421-1G APOSPC2421-250MG
4-Fluoro-2-(methoxymethyl)bromobenzene 98%
4-Fluoro-2-(methoxymethyl)bromobenzene
Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116323
CAS Number: 842167-67-3