1-[2-(Trifluoromethyl)phenyl]propylamine 98%
Supplier: Apollo Scientific
Synonyms: 2-(1-Aminoprop-1-yl)benzotrifluoride, α-Ethyl-2-(trifluoromethyl)benzylamine
PC2542-1G PC2542-250MG
APOSPC2542-1GEA 654 EUR
APOSPC2542-1G APOSPC2542-250MG
1-[2-(Trifluoromethyl)phenyl]propylamine 98%
1-[2-(Trifluoromethyl)phenyl]propylamine
Formula: C₁₀H₁₂F₃N
MW: 203.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784288
CAS Number: 473732-54-6