2',6'-Dichloro-4'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,6-Dichloro-4-(trifluoromethyl)phenyl)ethanone
PC2552E-250MG
APOSPC2552E-250MGEA 26.1 EUR
APOSPC2552E-250MG
2',6'-Dichloro-4'-(trifluoromethyl)acetophenone 97%
2',6'-Dichloro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₅Cl₂F₃O
MW: 257.037 g/mol
Boiling Pt: 58 °C (0.6 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00221015
CAS Number: 175205-88-6