3-Chloro-4,5-difluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC2592-1G PC2592-25G PC2592-5G
APOSPC2592-1GEA 0 EUR
APOSPC2592-1G APOSPC2592-25G APOSPC2592-5G
3-Chloro-4,5-difluorobenzoic acid 97%
3-Chloro-4,5-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
MW: 192.547 g/mol
Melting Pt: 130…133 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972707
CAS Number: 150444-95-4