5-Bromo-2-fluorophenyl methyl sulphone 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromo-1-fluoro-2-(methylsulphonyl)benzene
PC2624-1G
PC2624-5G
APOSPC2624-1GEA
116
EUR
APOSPC2624-1G
APOSPC2624-5G
5-Bromo-2-fluorophenyl methyl sulphone 99%
5-Bromo-2-fluorophenyl methyl sulphone
Formula:
C₇H₆BrFO₂S MW: 253.089 g/mol Melting Pt: 95…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458078 CAS Number: 914636-38-7 |