3-(Trifluoromethoxy)benzyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Chloromethyl)phenyl trifluoromethyl ether, 1-(Chloromethyl)-3-(trifluoromethoxy)benzene
PC2637-1G PC2637-5G PC2637-25G
APOSPC2637-1GEA 128 EUR
APOSPC2637-1G APOSPC2637-5G APOSPC2637-25G
3-(Trifluoromethoxy)benzyl chloride
3-(Trifluoromethoxy)benzyl chloride
Formula: C₈H₆ClF₃O
MW: 210.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458087
CAS Number: 89807-43-2
UN: 1760
ADR: 8 III