2-Chloro-1-iodo-4-(trifluoromethoxy)benzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-4-(trifluoromethoxy)iodobenzene, 3-Chloro-4-iodophenyl trifluoromethyl ether, 3-Chloro-4-iodo-α,α,α-trifluoroanisole
PC2685-1G PC2685-25G PC2685-5G
APOSPC2685-1GEA 21.9 EUR
APOSPC2685-1G APOSPC2685-25G APOSPC2685-5G
2-Chloro-1-iodo-4-(trifluoromethoxy)benzene 98%
2-Chloro-1-iodo-4-(trifluoromethoxy)benzene
Formula: C₇H₃ClF₃IO
MW: 322.451 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458089
CAS Number: 345226-19-9