2-Amino-6-bromo-4-fluorophenol 95%
Supplier: Apollo Scientific
Warning
PC2774-1G PC2774-250MG PC2774-5G
APOSPC2774-1GEA 328 EUR
APOSPC2774-1G APOSPC2774-250MG APOSPC2774-5G
2-Amino-6-bromo-4-fluorophenol 95%
2-Amino-6-bromo-4-fluorophenol
Formula: C₆H₅BrFNO
MW: 206.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094183
CAS Number: 502496-32-4