2-Nitro-4-(trifluoromethoxy)phenol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-5-(trifluoromethoxy)nitrobenzene
PC2782-1G PC2782-5G PC2782-10G
APOSPC2782-1GEA 132 EUR
APOSPC2782-1G APOSPC2782-5G APOSPC2782-10G
2-Nitro-4-(trifluoromethoxy)phenol
2-Nitro-4-(trifluoromethoxy)phenol
Formula: C₇H₄F₃NO₄
MW: 223.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094188
CAS Number: 129644-56-0